Human Albumin ELISA Kit
Leverantör: Abcam
Human Albumin ELISA kit is a single-wash, 90-minute SimpleStep ELISA® used to quantify human and cynomolgus monkey albumin (ALB) with a sensitivity of 0,1 ng/ml. The assay employs a simple Mix-Wash-Read protocol with just one incubation and wash step. - Colorimetric sandwich ELISA - 450 nm readout, compatible with any standard plate reader - Different formats for different needs: -384-well format available for high-throughput applications - Broad sample compatibility – works with serum, plasma (heparin, citrate, EDTA), and cell culture supernatants - Cited in over 15 publications.