Human Transferrin ELISA Kit
Leverantör: Abcam
AB187391-96 AB187391-1X384
ABCAAB187391-96EA 9725 SEK
ABCAAB187391-96 ABCAAB187391-1X384
Human Transferrin ELISA Kit
Analyser ELISAs
Human Transferrin ELISA Kit is a single-wash 90-min Simplestep used to quantify Human Transferrin with a sensitivity of 0,39 ng/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 384-well plate for higher throughput.