Ammonium 1-pyrrolidinecarbodithioate 98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Pyrrolidinedithiocarbamate ammonium salt, 1-Pyrrolidinecarbodithioic acid, ammonium salt, Ammonium pyrrolidine-1-carbodithioate, Pyrrolidine-1-Dithiocarboxylic Acid Ammonium Salt, APCD
162050250 162051000 162055000
ACRO162050250EA 825 SEK
ACRO162050250 ACRO162051000 ACRO162055000
Ammonium 1-pyrrolidinecarbodithioate 98%
Ammonium 1-pyrrolidinecarbodithioate
Formel: C₅H₁₂N₂S₂
MW: 146.3 · 18.0 g/mol
Smältpunkt: 150…155 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00012720
CAS nummer: 5108-96-3
EINECS: 225-834-4
Order Now
Specifikation testresultat
Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥96.0 %