Methyl-2-oxocyclopentanecarboxylate ≥97%
Leverantör: Thermo Fisher Scientific
Synonyms: Methyl 2-cyclopentanonecarboxylate
B25256.06 B25256.14 B25256.22
B25256.06EA 745 SEK
B25256.06 B25256.14 B25256.22
Methyl-2-oxocyclopentanecarboxylate ≥97%
Methyl-2-oxocyclopentanecarboxylate
Purity: 97% CAS No.: 10472-24-9 Molecular Formula: C7H10O3 Formula Weight: 142.16 Flash Point: 110°(230°F) Refractive Index: 1.4565 Boiling Point: 105-107°/20mm MDL No.: MFCD00001411
Formel: C₇H₁₀O₃
MW: 142.16 g/mol
Kokpunkt: 105…107 °C (20 mmHg)
Densitet: 1.145
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00001411
CAS nummer: 10472-24-9
EINECS: 233-962-7