1-Methyl-1,4-cyclohexadiene ≥97% stabilised
Leverantör: Thermo Scientific
Fara
Stabilization: Stabilised med BHT/Ionol
B20298.06 B20298.14
B20298.06EA 1350 SEK
B20298.06 B20298.14
1-Methyl-1,4-cyclohexadiene ≥97% stabilised
1-Methyl-1,4-cyclohexadiene
Stabilized with 0.01% BHT
Purity: 97% CAS No.: 4313-57-9 Molecular Formula: C7H10 Formula Weight: 94.16 Flash Point: 10°(50°F) Refractive Index: 1.4710 Odour: Characteristic Boiling Point: 114-115° Beilstein Reference: 1560168 UN No.: 3295 MDL No.: MFCD00001538
Formel: C₇H₁₀
MW: 94,16 g/mol
Kokpunkt: 114…115 °C
Densitet: 0,838
Flampunkt: 10 °C (50 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00001538
CAS nummer: 4313-57-9
EINECS: 224-329-6
ADR: 3,II