5-Amino-4,6-dichloro-2-methylpyrimidine ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 4,6-dichloro-2-methylpyrimidin-5-amine

A1946-25G A1946-5G
TCIAA1946-25GEA 4825 SEK
TCIAA1946-25G TCIAA1946-5G
5-Amino-4,6-dichloro-2-methylpyrimidine ≥98.0%
5-Amino-4,6-dichloro-2-methylpyrimidine
Formel: C₅H₅Cl₂N₃
MW: 178,0200 g/mol
MDL Number: MFCD00194053
CAS nummer: 39906-04-2
EINECS: 419-110-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=143e633880ab325ebc69251515bb9cc2fcbafdcb&vid=143e633880ab325ebc69251515bb9cc2fcbafdcb&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 71.0 to 75.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR