Chromone-2-carboxylic acid 97%
Leverantör: Thermo Scientific
Varning
Synonyms: 4-Oxo-4H-1-benzopyran-2-carboxylic acid, Chromocarb, 4-Oxo-4H-chromene-2-carboxylic acid
123880250 123880050
ACRO123880250EA 1750 SEK
ACRO123880250 ACRO123880050
Chromone-2-carboxylic acid 97%
Chromone-2-carboxylic acid
Formel: C₁₀H₆O₄
MW: 190,15 g/mol
Smältpunkt: 260 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00006838
CAS nummer: 4940-39-0
EINECS: 225-583-0
Order Now
Specifikation testresultat
Appearance White to beige to grey-brown Powder
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %
HPLC ≥96.0 %
NMR (1H-NMR) Consistent with structure