2-Cyclopentylcyclopentanone ≥99.0%

Leverantör: TCI EUROPE

Synonyms: [1,1'-Bi(cyclopentan)]-2-one

C0931-25ML C0931-500ML
TCIAC0931-25MLEA 485 SEK
TCIAC0931-25ML TCIAC0931-500ML
2-Cyclopentylcyclopentanone ≥99.0%
2-Cyclopentylcyclopentanone
Formel: C₁₀H₁₆O
MW: 152,24 g/mol
Kokpunkt: 233°C
Flampunkt: 104°C
MDL Number: MFCD00051419
CAS nummer: 4884-24-6
EINECS: 225-495-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8819d989aee5205374adc7a0d7a17426775c7998&vid=8819d989aee5205374adc7a0d7a17426775c7998&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR