Methyl-4-(bromomethyl)benzoate 98%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Methyl 4-(bromomethyl)benzoate
306100050 306100250 306101000
ACRO306100050EA 1175 SEK
ACRO306100050 ACRO306100250 ACRO306101000
Methyl-4-(bromomethyl)benzoate 98%
Methyl-4-(bromomethyl)benzoate
Formel: C₉H₉BrO₂
MW: 229.07 g/mol
Kokpunkt: 130…135 °C (2 mmHg)
Smältpunkt: 53…58 °C
Storage Temperature: Omgivande
MDL Number: MFCD00032453
CAS nummer: 2417-72-3
UN: 3261
ADR: 8 II
Order Now
Specifikation testresultat
Appearance White to yellow Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 52 to 57 °C
GC ≥97.5 %