3-Ethoxy-4-methoxybenzaldehyde ≥99%
Leverantör: Thermo Scientific
Synonyms: Apremilast Impurity 28
B24762.06 B24762.14 B24762.22
B24762.06EA 1175 SEK
B24762.06 B24762.14 B24762.22
3-Ethoxy-4-methoxybenzaldehyde ≥99%
3-Ethoxy-4-methoxybenzaldehyde
Purity: 99% CAS No.: 1131-52-8 Molecular Formula: C10H12O3 Formula Weight: 180.20 Melting Point: 51-53° MDL No.: MFCD00010128
Formel: C₁₀H₁₂O₃
MW: 180,20 g/mol
Smältpunkt: 51…53 °C
Storage Temperature: Omgivande
MDL Number: MFCD00010128
CAS nummer: 1131-52-8