Lithiumbis(trimethylsilyl)amide 1.0 mol/l i tert-butylmetyleter, AcroSeal®
Leverantör: Thermo Scientific
Fara
Synonyms: Lithiumbis(triméthylsilyl)amide, HMDS-Li, N-Lithiohexamethyldisilazane, HMDS lithium salt
345671000 345678000
345671000.EA 2550 SEK
345671000. 345678000.
Lithiumbis(trimethylsilyl)amide 1.0 mol/l i tert-butylmetyleter, AcroSeal®
Lithiumbis(trimethylsilyl)amide
Formel: C₆H₁₈LiNSi₂
MW: 167,33 g/mol
Kokpunkt: 55…56 °C (760 mmHg)
Densitet: 0,8 g/cm³
Flampunkt: –28 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008261
CAS nummer: 4039-32-1
EINECS: 223-725-6
UN: 2924
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Clear to hazy yellow to brown Solution
Titration Active base 21 to 25 % (as LiNSi)