Dihydrojasmone ≥97%

Leverantör: Thermo Scientific
Varning

Synonyms: 3-Methyl-2-pentylcyclopent-2-en-1-one

A19298.14 A19298.22
A19298.14EA 600 SEK
A19298.14 A19298.22
Dihydrojasmone ≥97%
Dihydrojasmone

Purity: 97% CAS No.: 1128-08-1 Molecular Formula: C11H18O Formula Weight: 166.26 Flash Point: 121°(250°F) Refractive Index: 1.479 Boiling Point: 120-121°/12mm Beilstein Reference: 1906471 MDL No.: MFCD00036480

Formel: C₁₁H₁₈O
MW: 166,26 g/mol
Kokpunkt: 120…121 °C (12 mmHg)
Densitet: 0,91
Flampunkt: 121 °C (250 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00036480
CAS nummer: 1128-08-1
EINECS: 214-434-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR