1,3,5,5-Tetramethyl-1,3-cyclohexadiene ≥92.0%
Leverantör: TCI EUROPE
|
Varning
|
T1718-1ML
T1718-5ML
TCIAT1718-1MLEA
615
SEK
TCIAT1718-1ML
TCIAT1718-5ML
1,3,5,5-Tetramethyl-1,3-cyclohexadiene ≥92.0%
1,3,5,5-Tetramethyl-1,3-cyclohexadiene
Formel:
C₁₀H₁₆ MW: 136,24 g/mol Flampunkt: 52°C |
MDL Number:
MFCD00045518 CAS nummer: 4724-89-4 UN: 3295 ADR: 3,III |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 92.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...