3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxylic acid ≥99%
Leverantör: Thermo Fisher Scientific
Varning
L10092.09 L10092.18
L10092.09EA 1500 SEK
L10092.09 L10092.18
3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxylic acid ≥99%
3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxylic acid
Purity: 99% CAS No.: 3919-74-2 Molecular Formula: C11H7ClFNO3 Formula Weight: 255.63 Melting Point: 201-203° Beilstein Reference: 806523
Formel: C₁₁H₇ClFNO₃
MW: 255.63 g/mol
Smältpunkt: 201…203 °C
Storage Temperature: Omgivande
MDL Number: MFCD00055659
CAS nummer: 3919-74-2
EINECS: 223-486-8