3-(2-Chlorophenyl)-5-methylisoxazole-4-carboxylic acid

Leverantör: Thermo Scientific

SB01885EA
MAYBSB01885EAEA 0 SEK
MAYBSB01885EA
3-(2-Chlorophenyl)-5-methylisoxazole-4-carboxylic acid
3-(2-Chlorophenyl)-5-methylisoxazole-4-carboxylic acid
Formel: C₁₁H₈ClNO₃
MW: 237,64 g/mol
MDL Number: MFCD00020813
CAS nummer: 23598-72-3
EINECS: 245-770-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR