3-(2-Chlorophenyl)-5-methylisoxazole-4-carboxylic acid ≥98%
Leverantör: Thermo Scientific
Varning
L06240.18 L06240.30 L06240.09
L06240.18EA 1450 SEK
L06240.18 L06240.30 L06240.09
3-(2-Chlorophenyl)-5-methylisoxazole-4-carboxylic acid ≥98%
3-(2-Chlorophenyl)-5-methylisoxazole-4-carboxylic acid
Purity: 98+% CAS No.: 23598-72-3 Molecular Formula: C11H8ClNO3 Formula Weight: 237.64 Melting Point: 187-192° Beilstein Reference: 791928
Formel: C₁₁H₈ClNO₃
MW: 237,64 g/mol
Smältpunkt: 187…192 °C
Storage Temperature: Omgivande
MDL Number: MFCD00020813
CAS nummer: 23598-72-3
EINECS: 245-770-0