Nicotinoyl chloride hydrochloride ≥98%
Leverantör: Thermo Scientific
Fara
A15317.09 A15317.18 A15317.30
A15317.09EA 720 SEK
A15317.09 A15317.18 A15317.30
Nicotinoyl chloride hydrochloride ≥98%
Nicotinoyl chloride hydrochloride
Purity: 98% CAS No.: 20260-53-1 Molecular Formula: C6H4ClNO.HCl Formula Weight: 178.02 Melting Point: 149-155° Beilstein Reference: 3655772 UN No.: 3261 MDL No.: MFCD00012820
Formel: C₆H₅Cl₂NO
MW: 178,02 g/mol
Smältpunkt: 149…155 °C
Storage Temperature: Omgivande
MDL Number: MFCD00012820
CAS nummer: 20260-53-1
EINECS: 243-648-1
UN: 3261
ADR: 8,II