4-(4-Bromophenoxy)benzaldehyde ≥97%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 4-(4-Bromo-phenoxy)benzaldehyde
H31702.03 H31702.06
H31702.03EA 1550 SEK
H31702.03 H31702.06
4-(4-Bromophenoxy)benzaldehyde ≥97%
4-(4-Bromophenoxy)benzaldehyde
Purity: 97% CAS No.: 69240-56-8 Molecular Formula: C13H9BrO2 Formula Weight: 277.12 Melting Point: 69-73° UN No.: 3077 MDL No.: MFCD08276811
Formel: C₁₃H₉BrO₂
MW: 277.12 g/mol
Smältpunkt: 69…73 °C
Storage Temperature: Omgivande
MDL Number: MFCD08276811
CAS nummer: 69240-56-8
ADR: 9 III