Ethyl-2-hydroxyisobutyrate ≥98%
Leverantör: Thermo Scientific
Varning
B23592.14 B23592.22 B23592.36
B23592.14EA 915 SEK
B23592.14 B23592.22 B23592.36
Ethyl-2-hydroxyisobutyrate ≥98%
Ethyl-2-hydroxyisobutyrate
Purity: 98% CAS No.: 80-55-7 Molecular Formula: C6H12O3 Formula Weight: 132.16 Flash Point: 44°(111°F) Refractive Index: 1.4080 Boiling Point: 149-150° Beilstein Reference: 878308 UN No.: 3272 MDL No.: MFCD00004458
Formel: C₆H₁₂O₃
MW: 132,16 g/mol
Kokpunkt: 149…150 °C
Densitet: 0,980
Flampunkt: 44 °C (111 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00004458
CAS nummer: 80-55-7
EINECS: 201-290-3
UN: 3272
ADR: 3,III