2-Bromoisobutyryl bromide ≥98.0%
Leverantör: TCI
|
Fara
|
Synonyms:
2-Bromo-2-methylpropionyl bromide
B0607-25G
B0607-500G
B0607-100G
TCIAB0607-25GEA
215
SEK
TCIAB0607-25G
TCIAB0607-500G
TCIAB0607-100G
2-Bromoisobutyryl bromide ≥98.0%
2-Bromoisobutyryl bromide
Formel:
C₄H₆Br₂O Kokpunkt: 164°C |
MDL Number:
MFCD00000122 CAS nummer: 20769-85-1 EINECS: 244-017-3 UN: 3265 ADR: 8,III |
|
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...