Manganese(II) nitrate 50% (w/w) i vattenlösning
Leverantör: Thermo Scientific
Fara
Synonyms: Manganese dinitrate, Manganous nitrate
33340.36 33340.A3
33340.36EA 1300 SEK
33340.36 33340.A3
Manganese(II) nitrate 50% (w/w) i vattenlösning
Manganese(II) nitrate
CAS No.: 10377-66-9 Molecular Formula: MnN2O6 Formula Weight: 178.95 Physical Form: Liquid Appearance: Pink Odour: Amine-like pH: 1.0 UN No.: 3093 MDL No.: MFCD00011112
Formel: Mn(NO₃)₂
MW: 178,95 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00011112
CAS nummer: 10377-66-9
EINECS: 233-828-8
UN: 3093
ADR: 8,II