6-Bromo-3-formylchromone ≥97.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
6-Bromo-4-oxo-4H-chromene-3-carbaldehyde
B2183-25G
B2183-5G
TCIAB2183-25GEA
2750
SEK
TCIAB2183-25G
TCIAB2183-5G
6-Bromo-3-formylchromone ≥97.0%
6-Bromo-3-formylchromone
Formel:
C₁₀H₅BrO₃ MW: 253,05 g/mol |
MDL Number:
MFCD00191849 CAS nummer: 52817-12-6 |
Specifikation testresultat
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 190.0 to 193.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...