(5-Bromo-2-pyridinyl)methanol ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE

Synonyms: 5-Bromo-2-(hydroxymethyl)pyridine, 5-Bromo-2-hydroxymethylpyridine

B3492-1G B3492-5G
TCIAB3492-1GEA 895 SEK
TCIAB3492-1G TCIAB3492-5G
(5-Bromo-2-pyridinyl)methanol ≥98.0% (by GC, titration analysis)
(5-Bromo-2-pyridinyl)methanol
Formel: C₆H₆BrNO
MW: 188,0200 g/mol
Kokpunkt: 120°C
MDL Number: MFCD04035597
CAS nummer: 88139-91-7
EINECS: 476-710-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea24bb0b276cc502aabc79ef687214690cc9b352&vid=ea24bb0b276cc502aabc79ef687214690cc9b352&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 59.0 to 63.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR