3-Quinolineboronic acid (contains varying amounts of Anhydride)

Leverantör: TCI EUROPE
Varning

Synonyms: Quinoline-3-boronic acid, Quinolin-3-ylboronic acid

Q0080-1G Q0080-5G
TCIAQ0080-1GEA 630 SEK
TCIAQ0080-1G TCIAQ0080-5G
3-Quinolineboronic acid (contains varying amounts of Anhydride)
3-Quinolineboronic acid
Formel: C₉H₈BNO₂
MW: 172,98 g/mol
MDL Number: MFCD02183527
CAS nummer: 191162-39-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32ddf34bc8abce708acf74e97145db85039f9910&vid=32ddf34bc8abce708acf74e97145db85039f9910&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Dark green powder to crystal
Purity (with Total Nitrogen) 97.0 to 112.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR