Ethyl-N-acetylglycinate(Ac-Gly-OEt) ≥98.0%

Leverantör: TCI EUROPE

Synonyms: Ac-Gly-OEt

A0094-1G A0094-25G
TCIAA0094-1GEA 215 SEK
TCIAA0094-1G TCIAA0094-25G
Ethyl-N-acetylglycinate(Ac-Gly-OEt) ≥98.0%
Ethyl-N-acetylglycinate(Ac-Gly-OEt)
Formel: C₆H₁₁NO₃
MW: 145,1 g/mol
Kokpunkt: 143°C
MDL Number: MFCD00009173
CAS nummer: 1906-82-7
EINECS: 217-608-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78f459b4a0f8ef3a266892347b713cf83d4c0f35&vid=78f459b4a0f8ef3a266892347b713cf83d4c0f35&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Melting point 46.0 to 50.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR