4-(Methoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)

Leverantör: TCI
Varning

Synonyms: (4-(Methoxycarbonyl)phenyl)boronic acid, 4-(Methoxycarbonyl)benzeneboronic acid

M1907-1G M1907-25G M1907-5G
TCIAM1907-1GEA 270 SEK
TCIAM1907-1G TCIAM1907-25G TCIAM1907-5G
4-(Methoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)
4-(Methoxycarbonyl)phenylboronic acid
Formel: C₈H₉BO₄
Smältpunkt: 234°C
MDL Number: MFCD01632203
CAS nummer: 99768-12-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38404ded914a202ba8518c23fa56e6bcc7b17f66&vid=38404ded914a202ba8518c23fa56e6bcc7b17f66&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) 96.0 to 111.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR