4-(Octyloxy)phenol ≥98.0%
Leverantör: TCI
|
Varning
|
O0140-25G
O0140-5G
TCIAO0140-25GEA
2525
SEK
TCIAO0140-25G
TCIAO0140-5G
4-(Octyloxy)phenol ≥98.0%
4-(Octyloxy)phenol
Formel:
C₁₄H₂₂O₂ Kokpunkt: 127°C |
MDL Number:
MFCD00045779 CAS nummer: 3780-50-5 EINECS: 223-243-6 |
|
Specifikation testresultat
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 59.0 to 62.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...