Diethyl azodicarboxylate 40% ∼2.2 mol/l I toluen
Leverantör: TCI EUROPE
|
Fara
|
A0705-25G
A0705-250G
A0705-100G
TCIAA0705-25GEA
765
SEK
TCIAA0705-25G
TCIAA0705-250G
TCIAA0705-100G
Diethyl azodicarboxylate 40% ∼2.2 mol/l I toluen
Diethyl azodicarboxylate
Formel:
C₆H₁₀N₂O₄ MW: 174,16 g/mol Flampunkt: 41°C Storage Temperature: Kylskåp |
MDL Number:
MFCD00009103 CAS nummer: 1972-28-7 EINECS: 217-821-7 UN: 1993 ADR: 3,II |
Specifikation testresultat
Appearance | Orange to Brown clear liquid |
Concentration (Iodometric titration) | 40.0 to 44.0 w/w% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...