Malonyl dichloride ≥97.0%

Leverantör: TCI EUROPE
Fara

M0882-10G M0882-25G
TCIAM0882-10GEA 645 SEK
TCIAM0882-10G TCIAM0882-25G
Malonyl dichloride ≥97.0%
Malonyl dichloride
Formel: C₃H₂Cl₂O₂
MW: 140,95 g/mol
Kokpunkt: 60°C
Flampunkt: 47°C
Storage Temperature: Kylskåp
MDL Number: MFCD00000735
CAS nummer: 1663-67-8
EINECS: 216-772-9
UN: 2920
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3bb9377b4b106f10d270d5e091c1e97fbfc1a8cf&vid=3bb9377b4b106f10d270d5e091c1e97fbfc1a8cf&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Brown clear liquid
Purity (Argentmetric Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR