2-Imidazolidinethione ≥98.0%

Leverantör: TCI
Fara

Synonyms: 2-Mercaptoimidazoline, Ethlenethiourea, Ethylenethiourea, 2-Imidazolidinethione

I0004-25G I0004-500G
TCIAI0004-25GEA 215 SEK
TCIAI0004-25G TCIAI0004-500G
2-Imidazolidinethione ≥98.0%
2-Imidazolidinethione
Formel: C₃H₆N₂S
MW: 102.16 g/mol
MDL Number: MFCD00005276
CAS nummer: 96-45-7
EINECS: 202-506-9
Merck Index: 14,03803
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b36e27c417b83e52c55c85a14904af8e0180863&vid=4b36e27c417b83e52c55c85a14904af8e0180863&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Iodometric Titration) min. 98.0 %
Melting point 197.0 to 202.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR