3-Bromopyridine ≥98.0% stabilised
Leverantör: TCI EUROPE
|
Varning
|
Stabilization:
Stabilised with copper chip
B0651-25G
B0651-100G
TCIAB0651-25GEA
405
SEK
TCIAB0651-25G
TCIAB0651-100G
3-Bromopyridine ≥98.0% stabilised
3-Bromopyridine
Formel:
C₅H₄BrN MW: 158 g/mol Kokpunkt: 178°C Smältpunkt: –27 °C Densitet: 1,62 g/cm³ (20 °C) Flampunkt: 52°C Storage Temperature: Omgivande |
MDL Number:
MFCD00006373 CAS nummer: 626-55-1 EINECS: 210-952-0 UN: 1993 ADR: 3,III |
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...