Methacrolein ≥90.0% stabilised
Leverantör: TCI EUROPE
Fara
Synonyms: Acrolein
Stabilization: med hydrokinon
M0078-25ML M0078-500ML M0078-100ML
TCIAM0078-25MLEA 725 SEK
TCIAM0078-25ML TCIAM0078-500ML TCIAM0078-100ML
Methacrolein ≥90.0% stabilised
Methacrolein
Formel: C₄H₆O
MW: 70,09 g/mol
Kokpunkt: 68°C
Smältpunkt: -81°C
Flampunkt: -15°C
Storage Temperature: Kylskåp
MDL Number: MFCD00006974
CAS nummer: 78-85-3
EINECS: 201-150-1
UN: 2396
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae12aa6c13ef5cb316c0bb7e8d07082f65307568&vid=ae12aa6c13ef5cb316c0bb7e8d07082f65307568&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 90.0 %