Sodium hexanitritocobaltate(III)
Leverantör: Thermo Scientific
Fara
Synonyms: Trinatriumhexanitritokobaltat
A11216.18 A11216.30
A11216.18EA 1075 SEK
A11216.18 A11216.30
Sodium hexanitritocobaltate(III)
Sodium hexanitritocobaltate(III)
CAS No.: 14649-73-1 Molecular Formula: CoN6Na3O12 Formula Weight: 403.94 Odour: Odourless Melting Point: 220° dec. Merck Reference: 14,8603 MDL No.: MFCD00003512
Formel: CoN₆Na₃O₁₂
MW: 403,94 g/mol
Smältpunkt: 220 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00003512
CAS nummer: 13600-98-1
EINECS: 237-077-7
UN: 1479
ADR: 5.1,II
Merck Index: 14,8603