4-Nitro-m-xylene ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: 2,4-Dimethyl-1-nitrobenzene, 1,3-Dimethyl-4-nitrobenzene

D0272-25G D0272-500G
TCIAD0272-25GEA 215 SEK
TCIAD0272-25G TCIAD0272-500G
4-Nitro-m-xylene ≥98.0%
4-Nitro-m-xylene
Formel: C₈H₉NO₂
MW: 151,16 g/mol
Kokpunkt: 245°C
Smältpunkt: 9 °C
Densitet: 1,135 g/cm³ (20 °C)
Flampunkt: 107°C
Storage Temperature: Omgivande
MDL Number: MFCD00007169
CAS nummer: 89-87-2
EINECS: 201-947-4
UN: 1665
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78dce9d16f2fd8af040adcba78517db0b8167471&vid=10daf84738173e7604fae428a67e0afdb6a9fdb9&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR