2-Nitro-m-xylene ≥99.0%

Leverantör: TCI
Fara

D0273-25G D0273-500G
TCIAD0273-25GEA 670 SEK
TCIAD0273-25G TCIAD0273-500G
2-Nitro-m-xylene ≥99.0%
2-Nitro-m-xylene
Formel: C₈H₉NO₂
Kokpunkt: 222°C
Smältpunkt: 14…16 °C
Densitet: 1,112 g/cm³ (20 °C)
Flampunkt: 87°C
Storage Temperature: Omgivande
MDL Number: MFCD00007181
CAS nummer: 81-20-9
EINECS: 201-333-6
UN: 1665
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1b71af59628a4a055d8569543b593d33c3d07fd4&vid=facbecef05edb078de1dadccbb89bb43f298b7ea&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White or Colorless to Yellow powder to lump to clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR