1,2-Diiodobenzene ≥98.0% stabilised
Leverantör: TCI EUROPE
Varning
Stabilization: Stabilised with copper chip
D0606-25G D0606-5G
TCIAD0606-25GEA 6750 SEK
TCIAD0606-25G TCIAD0606-5G
1,2-Diiodobenzene ≥98.0% stabilised
1,2-Diiodobenzene
Formel: C₆H₄I₂
MW: 329,91 g/mol
Kokpunkt: 152°C
Densitet: 2,76 g/cm³ (0 °C)
Storage Temperature: Kylskåp
MDL Number: MFCD00001038
CAS nummer: 615-42-9
EINECS: 210-425-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=60f555c076b2d2ba390417713d3855e93b2ef44e&vid=914579d64d3fe130f38f7f160e8dc571cfffff02&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White or Colorles to Yellow to Orange powder to lump to clear liquid
Purity (GC) min. 98.0 %