4-Pentyloxybenzaldehyde ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 4-(Pentyloxy)benzaldehyde, 4-n-Pentyloxybenzaldehyde, 4-Amyloxybenzaldehyde, 4-n-Amyloxybenzaldehyde

A0707-25ML A0707-500ML
TCIAA0707-25MLEA 550 SEK
TCIAA0707-25ML TCIAA0707-500ML
4-Pentyloxybenzaldehyde ≥98.0%
4-Pentyloxybenzaldehyde

Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: 4-Pentyloxybenzaldehyde

Formel: C₁₂H₁₆O₂
MW: 192,26 g/mol
Kokpunkt: 158°C
MDL Number: MFCD00014135
CAS nummer: 5736-91-4
EINECS: 227-250-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c71a0a4d97530cbe4534928d9ce16c0ed6aed36&vid=136f04c8f52f1a09c0dce546615a52ac2c84c635&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR