1-Bromo-3-iodobenzene ≥97.0% stabilised
Leverantör: TCI EUROPE
Varning
Synonyms: 3-Bromoiodobenzene
Stabilization: Stabilised with copper chip
B1236-25G B1236-5G B1236-100G
TCIAB1236-25GEA 590 SEK
TCIAB1236-25G TCIAB1236-5G TCIAB1236-100G
1-Bromo-3-iodobenzene ≥97.0% stabilised
1-Bromo-3-iodobenzene
Formel: C₆H₄BrI
MW: 282,91 g/mol
Kokpunkt: 120°C
Smältpunkt: –9,3 °C
Densitet: 2,21 g/cm³ (20 °C)
MDL Number: MFCD00001043
CAS nummer: 591-18-4
EINECS: 209-703-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a273dc48c24fa8a4c683be448b158fecbe8096d7&vid=1995e76313d2b9e54bc75a24a2782eba4ff61408&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Yellow clear liquid
Purity (GC) min. 97.0 %