2,2,2-Trifluoroethyl acrylate ≥98.0% stabilised
Leverantör: TCI
|
Fara
|
Synonyms:
2,2,2-Trifluoroethylacrylate, Acrylic acid 2,2,2-trifluoroethyl ester
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
A1152-25G
A1152-5G
TCIAA1152-25GEA
925
SEK
TCIAA1152-25G
TCIAA1152-5G
2,2,2-Trifluoroethyl acrylate ≥98.0% stabilised
2,2,2-Trifluoroethyl acrylate
Formel:
C₅H₅F₃O₂ Kokpunkt: 92°C Flampunkt: 13°C |
MDL Number:
MFCD00000444 CAS nummer: 407-47-6 EINECS: 206-987-6 UN: 3272 ADR: 3,II |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...