2-Piperidinoethanol ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
1-(2-Hydroxyethyl)piperidine, 1-Piperidineethanol
P0348-25G
P0348-500G
TCIAP0348-25GEA
215
SEK
TCIAP0348-25G
TCIAP0348-500G
2-Piperidinoethanol ≥98.0%
2-Piperidinoethanol
Formel:
C₇H₁₅NO MW: 129,2 g/mol Kokpunkt: 198°C Flampunkt: 73°C |
MDL Number:
MFCD00006512 CAS nummer: 3040-44-6 EINECS: 221-244-6 |
Specifikation testresultat
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...