1,4-Dibenzoylbutane ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 1,6-Diphenyl-1,6-butanedione, 1,6-Diphenylbutane-1,6-dione

D2773-25G D2773-5G
TCIAD2773-25GEA 9450 SEK
TCIAD2773-25G TCIAD2773-5G
1,4-Dibenzoylbutane ≥98.0%
1,4-Dibenzoylbutane
Formel: C₁₈H₁₈O₂
MW: 266,34 g/mol
MDL Number: MFCD00004761
CAS nummer: 3375-38-0
EINECS: 222-166-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=701893a830daf2dbf61834819162e38c1557f5b1&vid=701893a830daf2dbf61834819162e38c1557f5b1&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 105.0 to 109.0 deg-C
Solubility in hot Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR