1H,1H,10H,10H-Hexadecafluoro-1,10-decandiol ≥97.0%

Leverantör: TCI EUROPE

Synonyms: 1H,1H,10H,10H-Perfluoro-1,10-decandiol, 1H,1h,10h,10h-perfluoro-1,10-decanediol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-Hexadecafluoro-1,10-decandiol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-Hexadecafluoro-1,10-decanediol, 1H,1H,10H,10H-Perfluoro-1,10-decanediol, 1H,1H,10H,10H-Perfluorodecane-1,10-diol

H1233-1G H1233-5G
TCIAH1233-1GEA 270 SEK
TCIAH1233-1G TCIAH1233-5G
1H,1H,10H,10H-Hexadecafluoro-1,10-decandiol ≥97.0%
1H,1H,10H,10H-Hexadecafluoro-1,10-decandiol
Formel: C₁₀H₆F₁₆O₂
MW: 462,13 g/mol
Kokpunkt: 132°C
MDL Number: MFCD00192200
CAS nummer: 754-96-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=069e74d9e17074a24e2a76bd5f342dc55df2b02e&vid=069e74d9e17074a24e2a76bd5f342dc55df2b02e&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 136.0 to 140.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR