2-Jodpropan ≥99.0% stabilised
Leverantör: TCI EUROPE
Varning
Synonyms: Isopropyl iodide
Stabilization: Stabilised with copper chip
I0069-25G I0069-500G I0069-100G
TCIAI0069-25GEA 365 SEK
TCIAI0069-25G TCIAI0069-500G TCIAI0069-100G
2-Jodpropan ≥99.0% stabilised
2-Jodpropan
Formel: C₃H₇I
MW: 169,99 g/mol
Kokpunkt: 90°C
Smältpunkt: -90°C
Densitet: 1,7 g/cm³ (20 °C)
Flampunkt: 42°C
Storage Temperature: Omgivande
MDL Number: MFCD00001071
CAS nummer: 75-30-9
EINECS: 200-859-3
UN: 2392
ADR: 3,III
Merck Index: 14,05214
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec788505b6846ad10a477ebbf13f4c3d745af374&vid=d917a9e77fbba469f538fd38c0a44b97e8cbe9ee&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 99.0 %