1-Iodopentane ≥98.0% stabilised
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
1-Pentyl iodide
Stabilization:
Stabilised with copper chip
I0066-25ML
I0066-250ML
TCIAI0066-25MLEA
510
SEK
TCIAI0066-25ML
TCIAI0066-250ML
1-Iodopentane ≥98.0% stabilised
1-Iodopentane
Formel:
C₅H₁₁I MW: 198,05 g/mol Kokpunkt: 155°C Smältpunkt: –85,6 °C Densitet: 1,51 g/cm³ (20 °C) Flampunkt: 43°C Storage Temperature: Omgivande |
MDL Number:
MFCD00001100 CAS nummer: 628-17-1 EINECS: 211-030-0 UN: 1993 ADR: 3,III |
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...