1-Iodoperfluoropropane ≥95.0% stabilised

Leverantör: TCI EUROPE
Varning

Synonyms: Perfluoropropyl iodide, Heptafluoro-1-iodopropane, 1,1,2,2,3,3,3-Heptafluoro-1-iodopropane

Stabilization: Stabilised with copper chip

H0596-25G H0596-5G
TCIAH0596-25GEA 6000 SEK
TCIAH0596-25G TCIAH0596-5G
1-Iodoperfluoropropane ≥95.0% stabilised
1-Iodoperfluoropropane
Formel: C₃F₇I
MW: 295,93 g/mol
Kokpunkt: 41°C
Smältpunkt: –95 °C
Densitet: 2,1306 g/cm³ (0 °C)
Storage Temperature: Kylskåp
MDL Number: MFCD00001061
CAS nummer: 754-34-7
EINECS: 212-045-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=acbfc5cdc81ebccd76ad1e5fec1d939a63331cf1&vid=48b5cd064be3137afabc18d0a1b737e4abf7a790&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light red clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR