3-Chloro-1-iodopropane ≥98.0% stabilised
Leverantör: TCI
Varning
Stabilization: Stabilised with copper chip
C1037-25G C1037-5G
TCIAC1037-25GEA 1325 SEK
TCIAC1037-25G TCIAC1037-5G
3-Chloro-1-iodopropane ≥98.0% stabilised
3-Chloro-1-iodopropane
Formel: C₃H₆ClI
Kokpunkt: 171°C
Densitet: 1,904 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00001096
CAS nummer: 6940-76-7
EINECS: 230-088-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f5489bb7771481252b13f254615a77f3e156673&vid=6f5489bb7771481252b13f254615a77f3e156673&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %