4-(Methylthio)butanone ≥97.0%
Leverantör: TCI
M2031-25G
M2031-5G
TCIAM2031-25GEA
1150
SEK
TCIAM2031-25G
TCIAM2031-5G
4-(Methylthio)butanone ≥97.0%
4-(Methylthio)butanone
Formel:
C₅H₁₀OS Kokpunkt: 106°C Flampunkt: 72°C |
MDL Number:
MFCD00040029 CAS nummer: 34047-39-7 EINECS: 251-810-8 ADR: 9, |
Specifikation testresultat
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...