Acetoin ≥98.0% (May exist as crystalline dimer)

Leverantör: TCI
Varning

Synonyms: Acetyl methyl carbinol, 3-Hydroxybutanone, 3-Hydroxy-2-butanone

H0225-25G H0225-500G
TCIAH0225-25GEA 335 SEK
TCIAH0225-25G TCIAH0225-500G
Acetoin ≥98.0% (May exist as crystalline dimer)
Acetoin
Formel: C₄H₈O₂
Kokpunkt: 144°C
Smältpunkt: 20 °C
Densitet: 0,998 g/cm³ (20 °C)
Flampunkt: 46°C
Storage Temperature: Kylskåp
MDL Number: MFCD00004521
CAS nummer: 513-86-0
EINECS: 208-174-1
UN: 2621
ADR: 3,III
Merck Index: 14,00064
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ec11d2db1ea00b7885a9d51a04d2f25285ae314&vid=e9029a8ae14be8319ff9e0896ff914d6328edeb8&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR