4-Ethyl-2-methoxyphenol ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 5-Ethyl-2-hydroxyanisole, 1-Hydroxy-2-methoxy-4-ethyl Benzene, 4-Ethylguaiacol

E0353-25ML E0353-5ML
TCIAE0353-25MLEA 1600 SEK
TCIAE0353-25ML TCIAE0353-5ML
4-Ethyl-2-methoxyphenol ≥98.0%
4-Ethyl-2-methoxyphenol
Formel: C₉H₁₂O₂
MW: 152,19 g/mol
Kokpunkt: 235°C
Flampunkt: 108°C
MDL Number: MFCD00038714
CAS nummer: 2785-89-9
EINECS: 220-500-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0dc4564a3754fa4d791a2e07165d46a9fb35e178&vid=0dc4564a3754fa4d791a2e07165d46a9fb35e178&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR