2-Bromo-2-butene ≥98.0% stabilised
Leverantör: TCI EUROPE
Fara
Stabilization: Stabilised with copper chip
B1412-25G B1412-500G
TCIAB1412-25GEA 565 SEK
TCIAB1412-25G TCIAB1412-500G
2-Bromo-2-butene ≥98.0% stabilised
2-Bromo-2-butene
Formel: C₄H₇Br
MW: 135 g/mol
Kokpunkt: 82 °C (1013 hPa)
Densitet: 1,32 g/cm³ (20 °C)
Flampunkt: 18°C
Storage Temperature: Omgivande
MDL Number: MFCD00000141
CAS nummer: 13294-71-8
EINECS: 236-316-2
UN: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e02926de0d9da2bf93085f9fc587ddc21404b7e&vid=9589f19bc3b6f1188f18e5a134d85b30afdacb5b&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Purity (GC) min. 98.0 % (total of isomer)